missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COASY, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00080347-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MAINRFRLENDLEELALYQIQLLKDLRHTENEEDKVSSSSFRQRMLGNLLRPPYERPELPTCLYVIGLTGISGSGKSSIAQRLKGLGAFVIDSDHLGHRAYAPGGPAYQPVVEAFGTDILHKDGIINRKVLGSRVFGNKKQLKILTDIMWPIIAKLAREEMDRAVAEGKRVCVIDAAVLLEAGWQNLVHEVWTAVIPETEAVRRIVERDGLSEAAAQSRLQSQMSGQQLVEQSHVVLSTLWEPHITQRQVEKAWALLQKRIPKTHQALDSpecifications
| COASY | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_001042531.1 | |
| COASY | |
| COASY (NP_001035996.1, 1 a.a. ∼ 269 a.a) full-length human protein. | |
| RUO | |
| 80347 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Coenzyme A synthase | |
| DPCK/FLJ35179/NBP/PPAT/UKR1/pOV-2 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction