Learn More
COBLL1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98671MFV610
Product Code. 18504082
  • € 451.00 / 0.10mL
Description
COBLL1 Polyclonal antibody specifically detects COBLL1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
COBLL1
mFluor Violet 610 SE