Learn More
Invitrogen™ COBRA1 Recombinant Rabbit Monoclonal Antibody (24GB3740)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557112
 View more versions of this product
Product Code. 30276613
  • € 151.00 / 20µL
Description
COBRA1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
NELFB is a subunit of negative elongation factor (NELF), which also includes NELFA (WHSC2), either NELFC or NELFD (TH1L), and NELFE (RDBP), a heterodimer of SPT4 (SUPT4H1), to cause transcriptional pausing of RNA polymerase II (Narita et al., 2003).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
COBRA1
Recombinant Monoclonal
Unconjugated
NELFB
A730008L03Rik; AB041607; AI663983; Cobra1; cofactor of BRCA1; DKFZp586B0519; KIAA1182; MNCb-5210; Negative elongation factor B; negative elongation factor complex member B; negative elongation factor complex member B, Cobra1; negative elongation factor protein B; NELFB; NELF-B; ortholog of human cofactor of BRCA1 (COBRA1); RGD1307832; RP13-122B23.3
Rabbit
Affinity chromatography
RUO
25920, 311796, 58202
-20°C
Liquid
Western Blot, Immunocytochemistry
24GB3740
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8C4Y3, Q8WX92
NELFB
A synthesized peptide derived from human NELFB (aa 1-200).
20 μL
Primary
Human, Mouse, Rat