missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COCO/DAND5 Antibody (OTI3D4), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72457MFV450
This item is not returnable.
View return policy
Description
COCO/DAND5 Monoclonal antibody specifically detects COCO/DAND5 in Human samples. It is validated for Flow Cytometry
Specifications
| COCO/DAND5 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 199699 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OTI3D4 | |
| Flow Cytometry | |
| CER2CRL2, cerberus-like 2, Cerberus-like protein 2, cerl-2, CERL2, CKTSF1B3MGC126849, COCO, Cysteine knot superfamily 1, BMP antagonist 3, DAN domain family member 5, DAN domain family, member 5, DANTE, DTE, FLJ38607, GREM3cerberus 2, Gremlin-3, SP1 | |
| Human recombinant protein fragment corresponding to amino acids 23-189 of human DAND5(NP_689867) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction