missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CoCoA Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21437CL1
This item is not returnable.
View return policy
Description
CoCoA Polyclonal antibody specifically detects CoCoA in Human samples. It is validated for Immunoprecipitation,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| CoCoA | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| calcium binding and coiled-coil domain 1, calphoglin, Cocoa, Coiled-coil coactivator protein, coiled-coil leucine zipper coactivator 1, inorganic pyrophosphatase activator, KIAA1536calcium-binding and coiled-coil domain-containing protein 1, Sarcoma antigen NY-SAR-3 | |
| Produced in rabbits immunized with E. coli-derived Human CoCoA fragment | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 57658 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction