missing translation for 'onlineSavingsMsg'
Learn More

CoCoA Antibody, Novus Biologicals™

Product Code. p-200006020 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18617654 25 μg 25µL
18687674 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18617654 Supplier Novus Biologicals Supplier No. NBP32143725ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CoCoA Polyclonal antibody specifically detects CoCoA in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CoCoA
Applications Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5 uL/mg of lysate, Immunohistochemistry-Paraffin 1:200-1:1000
Formulation PBS, pH 7.0
Gene Alias calcium binding and coiled-coil domain 1, calphoglin, Cocoa, Coiled-coil coactivator protein, coiled-coil leucine zipper coactivator 1, inorganic pyrophosphatase activator, KIAA1536calcium-binding and coiled-coil domain-containing protein 1, Sarcoma antigen NY-SAR-3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CoCoA fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57658
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.