Learn More
Hach Lange™ COD Cuvette Test
Product Code. 10530323
Change view
Click to view available options
Range:
0 to 1000mg/L O2
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range Unit Size
10530323 0 to 1000mg/L O2 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10530323 Supplier Hach Lange™ Supplier No. LCI400
  • ONLINE EXCLUSIVE PRICE
    € 172.50
    € 163.42 / 24 tests
    Save € 9.08
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Certifications/Compliance ISO 15705
No. of Tests 24
Range 0 to 1000mg/L O2
Test Method Dichromate
Type COD Cuvette Test
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • CSB/COD/DCO
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • Germ cell mutagenicity Category 1B
  • Carcinogenicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH208-Contains potassium dichromate. May produce an allergic reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid ... %, Mercury(II) sulfate, Silver sulfate, potassium dichromate