missing translation for 'onlineSavingsMsg'
Learn More

Hach Pacific Pty Ltd COD Standard Solution, 300 mg/L as COD (NIST)

Product Code. 10239944
Change view
Click to view available options
Volume (Metric):
200 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10239944 200 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10239944 Supplier Hach Pacific Pty Ltd Supplier No. 1218629

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For accuracy checks in Chemical Oxygen Demand (COD) determinations.

    Specifications

  • Description: COD (NIST)
  • Parameter: COD
  • Platform: Solution
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 200 mL
For Use With (Equipment) Accuracy Checks in Chemical Oxygen Demand (COD) Determination
Type Solution
compliance-icons
Product Identifier
  • COD Standard Solution
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contain: Potassium hydrogen phthalate, copper sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.