Learn More
Coenzyme Q9, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Ubiquinone-9 |
| CAS | 303-97-9 |
| Molecular Formula | C54 H82 O4 |
| Quantity | 1 mg |
| Synonym | Ubiquinone Q9, Ubiquinone 9, CoQ9, NSC 226993, 2,3-dimethoxy-5-methyl-6-[(2E,6E,10E,14E,18E,22E,26E,30E)-3,7,11,15,19,23,27,31,35-nonamethyl-2,6,10,14,18,22,26,30,34-hexatriacontanonaen-1-yl]-2,5-Cyclohexadiene-1,4-dione, Coenzyme Q9, 2,3-dimethoxy-5-methyl-6-(3,7,11,15,19,23,27,31,35-nonamethyl-2,6,10,14,18,22,26,30,34-hexatriacontanonaenyl)-p-Benzoquinone, Ubidecarenone EP impurity D |
| InChI Formula | InChI=1S/C54H82O4/c1-40(2)22-14-23-41(3)24-15-25-42(4)26-16-27-43(5)28-17-29-44(6)30-18-31-45(7)32-19-33-46(8)34-20-35-47(9)36-21-37-48(10)38-39-50-49(11)51(55)53(57-12)54(58-13)52(50)56/h22,24,26,28,30,32,34,36,38H,14-21,23,25,27,29,31,33,35,37,39H2,1-13H3/b41-24+,42-26+,43-28+,44-30+,45-32+,46-34+,47-36+,48-38+ |
| SMILES | COC1=C(OC)C(=O)C(=C(C)C1=O)C\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CC\C=C(/C)\CCC=C(C)C |
| IUPAC Name | 2,3-dimethoxy-5-methyl-6-[(2E,6E,10E,14E,18E,22E,26E,30E)-3,7,11,15,19,23,27,31,35-nonamethylhexatriaconta-2,6,10,14,18,22,26,30,34-nonaenyl]cyclohexa-2,5-diene-1,4-dione |
| Molecular Weight (g/mol) | 795.23 |
| Formula Weight | 794.6213 |