missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cofilin Antibody (1C1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-21044F
This item is not returnable.
View return policy
Description
Cofilin Monoclonal antibody specifically detects Cofilin in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| Cofilin | |
| Monoclonal | |
| FITC | |
| NP_005498 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 1C1 | |
| PBS | |
| 18 kDa phosphoprotein, CFL, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, p18 | |
| Recombinant human Cofilin (1-166aa) purified from E. coli | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Neuronal Cell Markers | |
| 1072 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction