Learn More
COG1 Mouse, anti-Human, Clone: 2D8D11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68642-1-IG-20UL
 View more versions of this product
Product Code. 18047805
  • € 145.00 / 20µL
Description
The COG1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human COG1. This antibody recognizes human, rabbit antigen. The COG1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
COG1
2D8D11
CDG2G, COG complex subunit 1, COG1, DKFZp762L1710, KIAA1381, LDLB
20 μL
9382
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Rabbit
150 kDa