missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COG3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 292.00 - € 624.00
Specifications
| Antigen | COG3 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18406350
|
Novus Biologicals
NBP1-80772-25ul |
25ul |
€ 292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18217315
|
Novus Biologicals
NBP1-80772 |
0.1 mL |
€ 624.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COG3 Polyclonal specifically detects COG3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| COG3 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q96JB2 | |
| 83548 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SLTSQSLPIELTSVVPESTEDILLKGFTSLGMEEERIETAQQFFSWFAKLQTQMDQDEGTKYRQMRDYLSGFQEQCDAILNDVNSALQHLESLQKQYL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| COG complex subunit 3, component of oligomeric golgi complex 3p94, SEC34conserved oligomeric Golgi complex subunit 3, tethering factor SEC34, Vesicle-docking protein SEC34 homolog | |
| COG3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title