missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COG3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531824
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, mosue testis. Predicted reactivity: Mouse (95%), Rat (95%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a component of the conserved oligomeric Golgi (COG) complex which is composed of eight different subunits and is required for normal Golgi morphology and localization. Defects in the COG complex result in multiple deficiencies in protein glycosylation. The protein encoded by this gene is involved in ER-Golgi transport.
Specifications
| COG3 | |
| Polyclonal | |
| Unconjugated | |
| COG3 | |
| COG complex subunit 3; COG3; component of oligomeric golgi complex 2; component of oligomeric golgi complex 3; Conserved oligomeric Golgi complex subunit 3; E430004N23; p94; SEC34; tethering factor SEC34; vesicle-docking protein SEC34 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 338337, 83548 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8CI04, Q96JB2 | |
| COG3 | |
| Recombinant protein encompassing a sequence within the center region of human COG3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction