Learn More
COG6 Antibody (S09-6K8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19606-50ul
 View more versions of this product
Product Code. 18682854
  • € 421.00 / 50µL
Description
COG6 Monoclonal antibody specifically detects COG6 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
COG6
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse
Purified
Western Blot
S09-6K8
Unconjugated
COG complex subunit 6, component of oligomeric golgi complex 6KIAA1134COD2complexed with Dor1p 2, conserved oligomeric Golgi complex protein 6, conserved oligomeric Golgi complex subunit 6, DKFZp313D191
A synthetic peptide of human COG6 (Uniprot # Q9Y2V7)
50 μg
Golgi Apparatus Markers
57511
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.