missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COG7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17874580
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17874580 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17874580 Supplier Invitrogen™ Supplier No. PA5140066

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: GFQESILTDK KNSAKNPWQE YNYLQKDNPA EYASLMEILY TLKEKGSSNH NLLAAPRAAL TRLNQQAHQL AFDSVFLRIK QQLLLISKMD SWNTAGIG Highest antigen sequence indentity to the following orthologs: Rat - 90%, Mouse - 92%.

This gene encodes a WD repeat-containing protein that interacts with the Cul4-Ddb1 E3 ligase macromolecular complex. Multiple alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COG7
Applications Immunocytochemistry
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COG7
Gene Accession No. P83436
Gene Alias 5630400E24Rik; CDG2E; COG complex subunit 7; Cog7; Component of oligomeric Golgi complex 7; conserved oligomeric Golgi complex subunit 7; Gm167; UNQ3082/PRO10013
Gene Symbols COG7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COG7. Recombinant protein control fragment (Product #RP-108834).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91949
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.