missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COG8 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15928304
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15928304 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15928304 Supplier Invitrogen™ Supplier No. PA565559

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: AKVTKIILAFH RAEEAAFSSG EQELFVQFCT VFLEDLVPYL NRCLQVLFPP AQIAQTLGIP PTQLSKYGNL GHVNIGAIQE PL Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.

This gene encodes a protein that is a component of the conserved oligomeric Golgi (COG) complex, a multiprotein complex that plays a structural role in the Golgi apparatus, and is involved in intracellular membrane trafficking and glycoprotein modification. Mutations in this gene cause congenital disorder of glycosylation, type IIh, a disease that is characterized by under-glycosylated serum proteins, and whose symptoms include severe psychomotor retardation, failure to thrive, seizures, and dairy and wheat product intolerance.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COG8
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COG8
Gene Accession No. Q96MW5
Gene Alias BB235941; C87832; CDG2H; COG complex subunit 8; Cog8; Component of oligomeric Golgi complex 8; conserved oligomeric golgi complex component 8; conserved oligomeric Golgi complex protein 8; conserved oligomeric Golgi complex subunit 8; dependent on RIC1; DOR1; MNCb-5704
Gene Symbols COG8
Host Species Rabbit
Immunogen Recombinant Human COG8. Recombinant protein control fragment (Product #RP-104871).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84342
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.