missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565371
This item is not returnable.
View return policy
Description
Immunogen sequence: PSVIKIHTLVE SLKLSITDQQ LPMFIRIMQL GIALYYGEIG NFKEGEIEDL TCHNKDMLGN ITGSEDETR Highest antigen sequence identity to the following orthologs - mouse 83%, rat 27%.
COH1 (Cohen syndrome protein 1), also known as VPS13B (vacuolar protein sorting-associated protein 13B) or CHS1, belongs to the VPS13 family and may function in vesicle-mediated transport and sorting of proteins within the cell. COH1 is widely expressed and multiple alternatively spliced transcript variants have been observed. Mutations in this gene have been associated with Cohen syndrome. COH1 is a Golgi-localized peripheral membrane protein and plays a critical role in Golgi (re)assembly.
Specifications
| COH1 | |
| Polyclonal | |
| Unconjugated | |
| VPS13B | |
| 1810042B05Rik; 2310042E16Rik; 4732488H20; C330002D13Rik; C87206; CHS1; COH1; Cohen syndrome homolog 1; Cohen syndrome protein 1; Cohen syndrome protein 1 homolog; D230005K13; KIAA0532; mKIAA0532; mKIAA5032; vacuolar protein sorting 13 homolog B; vacuolar protein sorting 13B (yeast); vacuolar protein sorting-associated protein 13B; Vps13b | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 157680 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z7G8 | |
| VPS13B | |
| Recombinant Human COH1. Recombinant protein control fragment (Product #RP-104778). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction