missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL11A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577919
This item is not returnable.
View return policy
Description
Positive Control: human COLC11A2-transfected 293T cells (partial fragment) Predicted Reactivity: Mouse (89%), Rat (89%), Bovine (93%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes one of the two alpha chains of type XI collagen, a minor fibrillar collagen. It is located on chromosome 6 very close to but separate from the gene for retinoid X receptor beta. Type XI collagen is a heterotrimer but the third alpha chain is a post-translationally modified alpha 1 type II chain. Proteolytic processing of this type XI chain produces PARP, a proline/arginine-rich protein that is an amino terminal domain. Mutations in this gene are associated with type III Stickler syndrome, otospondylomegaepiphyseal dysplasia (OSMED syndrome), Weissenbacher-Zweymuller syndrome, autosomal dominant non-syndromic sensorineural type 13 deafness (DFNA13), and autosomal recessive non-syndromic sensorineural type 53 deafness (DFNB53). Alternative splicing results in multiple transcript variants. A related pseudogene is located nearby on chromosome 6.
Specifications
| COL11A2 | |
| Polyclonal | |
| Unconjugated | |
| COL11A2 | |
| COL11A2; Collagen alpha-2(XI) chain; collagen type XI alpha 2; collagen type XI alpha 2 chain; collagen XI-like; collagen, type XI, alpha 2; DAQB-79P13.8; DFNA13; DFNB53; FBCG2; HGNC:2187; HKE5; PARP; pro-a2 chain of collagen type XI; procollagen, type XI, alpha 2; putative collagen XI; STL3; type XI collagen alpha2 chain | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1302 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.76 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P13942 | |
| COL11A2 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human COL11A2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction