missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL15A1 Antibody (OTI1C5), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70422V
This item is not returnable.
View return policy
Description
COL15A1 Monoclonal specifically detects COL15A1 in Human samples. It is validated for Western Blot.
Specifications
| COL15A1 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| COL15A1 | |
| Full length recombinant protein of human COL15A1 (NP_001846) produced in HEK293T cells. | |
| 0.1 mL | |
| 1306.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1C5 | |
| Western Blot | |
| alpha-1 polypeptide, collagen alpha-1(XV) chain, collagen, type XV, alpha 1, FLJ38566 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction