missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COL19A1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15757821 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15757821 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15757821 Supplier Invitrogen™ Supplier No. PA559975

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: VTVRDKTEES CPILRIEGHQ LTYDNINKLE VSGFDLGDSF SLRRAFCESD KTCFKLGSAL LIRDTIKIFP KGLPEEYSVA AMFRVRRNAK KERWFLWQVL Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 70%.

Collagen Type XIX is an 1,142 amino acid protein encoded by the human gene COL19A1. Collagen Type XIX belongs to the fibril-associated collagens with interrupted helices (FACIT) family. Collagen Type XIX is thought to act as a cross-bridge between collagen fibrils and other extracellular matrix molecules. Collagen Type XIX is also believed to be involved in skeletal myogenesis in the developing esophagus. It may also play a role in organization of the pericellular matrix or the sphinteric smooth muscle. Collagen Type XIX is normally found as an oligomer, the subunits being disulfide-linked. Expression of Collagen Type XIX is mainly localized to vascular, neuronal, mesenchymal, and some epithelial basement membrane zones in umbilical cord.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL19A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene COL19A1
Gene Accession No. Q14993
Gene Alias a1 chain of type XIX collagen; col19a1; COL9A1L; collagen alpha 1 (Y) chain; collagen alpha-1(XIX) chain; Collagen alpha-1(Y) chain; collagen type XIX alpha 1 chain; collagen XIX, alpha-1 polypeptide; collagen, type XIX, alpha 1; D6S228E; procollagen, type XIX, alpha 1
Gene Symbols COL19A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human COL19A1. Recombinant protein control fragment (Product #RP-92408).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1310
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.