missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COL1A1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16313815
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16313815 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16313815 Supplier Invitrogen™ Supplier No. PA586949

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

The protein encoded by this gene is the pro-alpha1 chain of type I collagen whose triple helix comprises two alpha1 chains and one alpha2 chain. Type I is a fibril-forming collagen found in most connective tissues and is abundant in bone, cornea, dermis and tendon. Mutations in this gene are associated with osteogenesis imperfecta types I-IV, Ehlers-Danlos syndrome type VIIA, Ehlers-Danlos syndrome Classical type, Caffey Disease and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL1A1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene COL1A1
Gene Accession No. P02452, P02454, P11087
Gene Alias alpha-1 type 1 collagen; alpha-1 type I collagen; alpha1(I) procollagen; COL1A; Col1a1; Col1a-1; Cola1; Cola-1; COLIA1; Collagen 1; collagen alpha 1 chain type I; collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; Collagen I; collagen of skin, tendon and bone, alpha-1 chain; collagen type I alpha 1; collagen, type 1, alpha 1; collagen, type I, alpha 1; Collagen1A1; EDSC; Mov13; Mov-13; OI1; OI2; OI3; OI4; pro-alpha-1 collagen type 1; procollagen type I, alpha 1; procollagen, type 1, alpha 1; procollagen, type I, alpha 1; Type I Collagen; type I proalpha 1; type I procollagen alpha 1 chain; type I-alpha 1 collagen
Gene Symbols COL1A1
Host Species Rabbit
Immunogen A synthetic peptide of human COL1A1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1277, 12842, 29393
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.