missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COL4A1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 30534987
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30534987 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30534987 Supplier Invitrogen™ Supplier No. PA5149180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Collagen IV alpha-1 (COL4A1) encodes the major type IV alpha collagen chain of basement membranes. Like the other members of the type IV collagen gene family, this gene is organized in a head-to-head conformation with another type IV collagen gene so that each gene pair shares a common promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen COL4A1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene COL4A1
Gene Accession No. P02462
Gene Alias alpha 1 type IV collagen; alpha1(IV) collagen; arresten; Bru; BSVD; COL4A1; Col4a-1; COL4A1 NC1 domain; collagen alpha-1(IV) chain; collagen IV, alpha-1 polypeptide; collagen of basement membrane, alpha-1 chain; collagen type IV alpha 1 chain; collagen, type IV, alpha 1; Del(8)44H; Del(8)Bru44H; procollagen, type IV, alpha 1; RATOR; Raw; retinal anterior wiring; Svc
Gene Symbols COL4A1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Collagen IV alpha 1, Ser1441-Thr1669.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1282
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.