missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL4A3BP Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2114AF594
This item is not returnable.
View return policy
Description
COL4A3BP Polyclonal antibody specifically detects COL4A3BP in Human samples. It is validated for Western Blot
Specifications
| COL4A3BP | |
| Polyclonal | |
| Western Blot | |
| Ceramide transfer protein, CERTFLJ20597, CERTL, collagen type IV alpha-3-binding protein, collagen, type IV, alpha 3 (Goodpasture antigen) binding protein, Goodpasture antigen-binding protein, GPBPSTARD11ceramide transporter, hCERT, lipid-transfer protein CERTL, StARD11, StAR-related lipid transfer (START) domain containing 11, StAR-related lipid transfer protein 11, START domain containing 11, START domain-containing protein 11 | |
| The immunogen recognized by this antibody maps to a region between residue 300 and 350 of human CERTL(GPBP) using the numbering given in entry NP_005704.1 (GeneID 10087). | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Immune System Diseases, Immunology | |
| 10087 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction