missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ COL4A3BP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30933-25ug
This item is not returnable.
View return policy
Description
COL4A3BP Polyclonal antibody specifically detects COL4A3BP in Human samples. It is validated for Western Blot
Specifications
| COL4A3BP | |
| Polyclonal | |
| Western Blot : 1-5 μg/mL | |
| Ceramide transfer protein, CERTFLJ20597, CERTL, collagen type IV alpha-3-binding protein, collagen, type IV, alpha 3 (Goodpasture antigen) binding protein, Goodpasture antigen-binding protein, GPBPSTARD11ceramide transporter, hCERT, lipid-transfer protein CERTL, StARD11, StAR-related lipid transfer (START) domain containing 11, StAR-related lipid transfer protein 11, START domain containing 11, START domain-containing protein 11 | |
| Synthetic peptide derived from human COL4A3BP | |
| 25 μg | |
| Cell Biology, Cellular Markers, Immune System Diseases, Immunology | |
| 10087 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction