missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL4A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119041
This item is not returnable.
View return policy
Description
COL4A4 Polyclonal Antibody for Western Blot, ICC/IF
Collagens are highly conserved and are characterized by an uninterrupted Glycine-X-Y triplet repeat. Col1, the fibrillar collagen found in most connective tissues, is the only component of the collagen found in cartilage. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, and idiopathic osteoporosis. Col2, a fibrillar collagen found in cartilage and the vitreous humor of the eye. Mutations in this gene are associated with achondrogenesis, chondrodysplasia, early onset familial osteoarthritis, SED congenita, Langer-Saldino achondrogenesis, Kniest dysplasia. Col3 is a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I collagen. Mutations in this gene are associated with Ehlers-Danlos syndrome type IV, aortic and arterial aneurysms. Col4 is a major consitiuent of the basement membranes along with laminins and enactins. It is composed of alpha 1 IV chain and alpha 2 IV chain in 2:1 ratio. Col4 antibodies are useful in detecting the loss of parts of basement membrane in carcinomas. Col5 is found in tissues containing Col1 and appears to regulate the assembly of heterotypic fibers composed of both Col1 and Col5. Col5 is closely related to type XI collagen; mutations in this gene are associated with Ehlers-Danlos syndrome.
Specifications
| COL4A4 | |
| Polyclonal | |
| Unconjugated | |
| COL4A4 | |
| CA44; COL4A4; Collagen 4; Collagen alpha-4(IV) chain; Collagen IV, alpha-4 polypeptide; collagen of basement membrane, alpha-4 chain; collagen type IV alpha 4 chain; collagen, type IV, alpha 4 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 1286 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P53420 | |
| COL4A4 | |
| Recombinant protein Collagen Type IV Alpha 4. The antigen corresponds to amino acid range 1469-1690 of the target protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction