missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL5A2, Mouse anti-Human, Clone: 3G11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00001290-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: CTQNGQMYLNRDIWKPAPCQICVCDNGAILCDKIECQDVLDCADPVTPPGECCPVCSQTPGGGNTNFGRGRKGQKGEPGLVPVVSpecifications
| COL5A2 | |
| Monoclonal | |
| Unconjugated | |
| collagen, type V, alpha 2 | |
| MGC105115 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 3G11 | |
| In 1x PBS, pH 7.4 | |
| NM_000393 | |
| COL5A2 | |
| COL5A2 (NP_000384, 41 a.a. ∼ 124 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 1290 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction