missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COL9A1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92796-0.1ml
This item is not returnable.
View return policy
Description
COL9A1 Polyclonal antibody specifically detects COL9A1 in Rat samples. It is validated for Western Blot
Specifications
| COL9A1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000 | |
| alpha-1 polypeptide, collagen, type IX, alpha 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-270 of human COL9A1 (NP_001842.3). WASAAVKRRPRFPVNSNSNGGNELCPKIRIGQDDLPGFDLISQFQVDKAASRRAIQRVVGSATLQVAYKLGNNVDFRIPTRNLYPSGLPEEYSFLTTFRMTGSTLKKNWNIWQIQDSSGKEQVGIKINGQTQSVVFSYKGLDGSLQTAAFSNLSSLFDSQWHKIMIGVERSSATLFVDCNRIESLPIKPRGPIDIDGFAVLGKLADNPQVSVPFELQWMLIHCDPLRPRRETCHELPARITPSQTTDERGP | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 1297 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction