missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COL9A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584981
This item is not returnable.
View return policy
Description
Immunogen sequence: RGVPGQPGRQ GVEGRDATDQ HIVDVALKML Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 83%.
This gene encodes one of the three alpha chains of type IX collagen, the major collagen component of hyaline cartilage. Type IX collagen, a heterotrimeric molecule, is usually found in tissues containing type II collagen, a fibrillar collagen. This chain is unusual in that, unlike the other two type IX alpha chains, it contains a covalently attached glycosaminoglycan side chain. Mutations in this gene are associated with multiple epiphyseal dysplasia. [provided by RefSeq, Jul 2008].
Specifications
| COL9A2 | |
| Polyclonal | |
| Unconjugated | |
| COL9A2 | |
| AI427499; alpha 1 type IX collagen; alpha 2 type IX collagen; COL9A2; Col9a-2; Collagen alpha-2(IX) chain; collagen IX, alpha-2 polypeptide; collagen type IX alpha 2; collagen, type IX, alpha 2; DJ39G22.4; EDM2; MED; procollagen, type IX, alpha 2; STL5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1298 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14055 | |
| COL9A2 | |
| Recombinant protein corresponding to Human COL9A2. Recombinant protein control fragment (Product #RP-108653). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction