missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COLEC11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546802
This item is not returnable.
View return policy
Description
Peptide sequence: AQAGLARVFI GINDLEKEGA FVYSDHSPMR TFNKWRSGEP NNAYDEEDCV.
This gene encodes a member of the collectin family of C-type lectins that possess collagen-like sequences and carbohydrate recognition domains. Collectins are secreted proteins that play important roles in the innate immune system by binding to carbohydrate antigens on microorganisms, facilitating their recognition and removal. The encoded protein binds to multiple sugars with a preference for fucose and mannose. Mutations in this gene are a cause of 3MC syndrome-2. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| COLEC11 | |
| Polyclonal | |
| Unconjugated | |
| COLEC11 | |
| 1010001H16Rik; 3MC2; CLK1; CL-K1; CL-K1-I; CL-K1-II; CL-K1-IIa; CL-K1-IIb; Colec11; Collectin 11; Collectin K1; collectin kidney protein 1; collectin subfamily member 11; collectin sub-family member 11; Collectin11; collectin-11; hypothetical protein LOC492459; UNQ596/PRO1182; wu:fb82e09; zgc:103572 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 78989 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BWP8 | |
| COLEC11 | |
| Synthetic peptide directed towards the C-terminal of human COLEC11 (aa 194-243). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction