missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Colistin sulfate, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Colistin sulfate is a polypeptide antibiotic which inhibits gram-negative bacteria by binding to lipopolysaccharides and phospholipids in the outer cell membrane of gram-negative bacteria.
Specifications
Specifications
| Chemical Name or Material | Colistin sulfate |
| Physical Form | Solid |
| CAS | 1264-72-8 |
| Color | Off-White |
| Quantity | 500 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C52H98N16O13.2.5H2O4S |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Polymyxin E sulfate |
| Solubility Information | H2O : 50 mg/mL (35.70 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction