missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen I/III Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-64523AF488
This item is not returnable.
View return policy
Description
Collagen I/III Polyclonal antibody specifically detects Collagen I/III in Bovine samples. It is validated for ELISA, Immunofluorescence
Specifications
| Collagen I/III | |
| Polyclonal | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Alpha-1 type I collagen, Collagen 1/3, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, OI4, pro-alpha-1 collagen type 1 | |
| Bovine collagen type I and III from skin | |
| 0.25 mL | |
| Cell Biology, Cellular Markers, Extracellular Matrix | |
| 1277 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA, Immunofluorescence | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction