missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Collagen III Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586281
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
Collagen III is a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I Collagen. Mutations in th gene are associated with Ehlers Danlos syndrome type IV, and with aortic and arterial aneurysms. Although alternate transcripts have been detected for this gene, they are the result of mutations; these mutations alter splicing, often leading to the exclusion of multiple exons.
Specifications
| Collagen III | |
| Polyclonal | |
| Unconjugated | |
| Col3a1 | |
| alpha1 (III) collagen; alpha-1 collagen type III; alpha-1 type III collagen; alpha-1 type-3 collagen; AW550625; COL 3; COL3; COL3 alpha 1; COL3A1; Col3a-1; collagen; Collagen 3; collagen alpha 1 type III; collagen alpha-1(III) chain; collagen type III alpha 1 chain; collagen, fetal; collagen, type III, alpha 1; collagen, type III, alpha 1 (Ehlers-Danlos syndrome type IV, autosomal dominant); collagen, type III, alpha 1 chain, fibril forming; Collagen3; EDS4A; Ehlers-Danlos syndrome type IV, autosomal dominant; FLJ34534; MGC93704; minisatellite 10w detected by probe MMS10; mKIAA4231; MMS10-W; Ms10w; PIIINP; pro alpha 1 type III collagen; procollagen, type III, alpha 1; unknown | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1281, 12825, 84032 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P02461, P08121, P13941 | |
| Col3a1 | |
| Synthetic peptide, corresponding to amino acids 71-120 of Human COL3A1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction