missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Collagen I Polyclonal Antibody
GREENER_CHOICE

Product Code. 11571572
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11571572 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11571572 Supplier Invitrogen™ Supplier No. PA126204

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Some class-specific anti-collagens are specific for three-dimensional epitopes, resulting in diminished reactivity with denatured collagen or formalin-fixed, paraffin embedded tissues. This antibody reacts with most mammalian Type I collagens and has negligible cross-reactivity with Type II, III, IV, V or VI collagens. Cross-reactivity with other human serum proteins and non-collagen extracellular matrix proteins is neglible. Western blot: this preparation results in a native conformation of the protein. It is recommend not to add 2-ME or DTT in lysis buffer and not boiling the sample.

Collagen I is a Type I collagen with a triple helix structure comprised of two alpha-1 chains and one alpha-2 chain. Collagen I is a member of group I collagen (fibril-forming collagen) found in most connective tissues, and is abundant in bone, cornea, dermis and tendon. Mutations in the COL1A2 gene encoding the alpha-2 chain are associated with osteogenesis imperfecta types I-IV, Ehlers-Danlos syndrome type VIIB, recessive Ehlers-Danlos syndrome Classical type, idiopathic osteoporosis, and atypical Marfan syndrome. Symptoms associated with mutations in the COL1A2 gene which codes for the alpha-2 chain, however, tend to be less severe than mutations in the COL1A1 gene, reflecting the different role of alpha-2 chains in matrix integrity. Reciprocal translocations between chromosomes 17 and 22, where Collagen Type 1 genes and the gene for platelet derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen I
Applications Dot Blot, ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.01% sodium azide; pH 8.0
Gene COL1A1
Gene Accession No. P02452, P02453, P02454, P02465, P02466, P08123, P11087, Q01149
Gene Alias AA960264; AI325291; alpha 1 type I collagen; alpha 2 type I collagen; alpha 2 type I procollagen; alpha 2(I) collagen; alpha 2(I) procollagen; alpha 2(I)-collagen; alpha1 chain of type I collagen; alpha-1 collagen (I); alpha-1 type 1 collagen; alpha-1 type I collagen; alpha-1 type-1 collagen; alpha1(I) procollagen; alpha2 chain of type I collagen; alpha-2 type I collagen; alpha-2(1) collagen; alpha2(I) collagen; COL1; Col1a1; Col1a-1; COL1A1; matrix protein; Col1a2; Col1a-2; COLA; Cola1; Cola-1; Cola2; Cola-2; COLIA1; collagen; collagen 1 alpha 2 chain; collagen a2; collagen alpha 1 chain type I; collagen alpha 2(I) chain precursor; collagen alpha-1 chain; collagen alpha-1 chain precursor; collagen alpha-1(I) chain; collagen alpha-1(I) chain preproprotein; collagen alpha-2; collagen alpha-2 chain precursor; collagen alpha-2(I) chain; collagen COL1A2; collagen I alpha-2 polypeptide; collagen I, alpha-2 polypeptide; collagen of skin, tendon and bone, alpha-1 chain; collagen of skin, tendon and bone, alpha-2 chain; collagen type I alpha 1; collagen type I alpha 1 chain; collagen type I alpha 2 chain; collagen, type 1, alpha 1; collagen, type I, alpha 1; collagen, type I, alpha 2; EDSC; Mov13; Mov-13; OI1; OI2; OI3; OI4; oim; osteogenesis imperfecta; precollagen alpha-2; prepro-alpha-1 collagen type I; pro-alpha-1 collagen type 1; procollagen type I, alpha 1; procollagen, type 1, alpha 1; procollagen, type I, alpha 1; procollagen, type I, alpha 2; similar to pro alpha 1(I) collagen; Type I Collagen; type I collagen alpha 1 chain; type I collagen alpha1; type I collagen alpha-2 chain; type I collagen pre-pro-alpha1(I) chain; type I proalpha 1; type I procollagen; type I procollagen alpha 1 chain; type I-alpha 1 collagen
Gene Symbols COL1A1, COL1A2
Host Species Rabbit
Immunogen Collagen Type I from human and bovine placenta.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100759814, 100766269, 1277, 1278, 12842, 12843, 282187, 282188, 29393, 84352
Target Species Human, Mouse, Rat, Bovine, Hamster
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.