missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen III alpha 1/COL3A1 Antibody [Alexa Fluor™ 488], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-594AF488
This item is not returnable.
View return policy
Description
Collagen III alpha 1/COL3A1 Polyclonal antibody specifically detects Collagen III alpha 1/COL3A1 in Bovine,Feline,Human,Mouse,Rat,Sheep samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Simple Western,Immunocytochemistry/ Immunofluorescence
Specifications
| Collagen III alpha 1/COL3A1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Bovine, Feline, Human, Mouse, Rat, Sheep | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Simple Western, Immunocytochemistry/Immunofluorescence | |
| Alexa Fluor 488 | |
| alpha1 (III) collagen, alpha-1 type III collagen, COL3A1, Collagen 3, collagen alpha-1(III) chain, collagen, fetal, collagen, type III, alpha 1, Collagen-3, Collagen-III, collagenIII, EDS4A, EDSVASC, Ehlers-Danlos syndrome type IV, autosomal dominant, FLJ34534, PMGEDSV | |
| Collagen III alpha 1/COL3A1 from human and bovine placenta (Uniprot: P02461) | |
| 0.1 mL | |
| Cancer, Cell Biology, Extracellular Matrix | |
| 1281 | |
| Store at 4⪚C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction