missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Collagen III Monoclonal Antibody (C24)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA541569
This item is not returnable.
View return policy
Description
Collagen III Monoclonal Antibody for Western Blot
Collagen III is a fibrillar collagen that is found in extensible connective tissues such as skin, lung, and the vascular system, frequently in association with type I Collagen. Mutations in th gene are associated with Ehlers Danlos syndrome type IV, and with aortic and arterial aneurysms. Although alternate transcripts have been detected for this gene, they are the result of mutations; these mutations alter splicing, often leading to the exclusion of multiple exons.
Specifications
| Collagen III | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P13941 | |
| Col3a1 | |
| Recombinant protein Collagen Type III Alpha 1. The antigen corresponds to amino acid range 1171-1435 of the target protein. | |
| 200 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| C24 | |
| Unconjugated | |
| Col3a1 | |
| alpha1 (III) collagen; alpha-1 collagen type III; alpha-1 type III collagen; alpha-1 type-3 collagen; AW550625; COL 3; COL3; COL3 alpha 1; COL3A1; Col3a-1; collagen; Collagen 3; collagen alpha 1 type III; collagen alpha-1(III) chain; collagen type III alpha 1 chain; collagen, fetal; collagen, type III, alpha 1; collagen, type III, alpha 1 (Ehlers-Danlos syndrome type IV, autosomal dominant); collagen, type III, alpha 1 chain, fibril forming; Collagen3; EDS4A; Ehlers-Danlos syndrome type IV, autosomal dominant; FLJ34534; MGC93704; minisatellite 10w detected by probe MMS10; mKIAA4231; MMS10-W; Ms10w; PIIINP; pro alpha 1 type III collagen; procollagen, type III, alpha 1; unknown | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 84032 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction