missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV alpha5 Antibody (COL4A5/9396R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 279.00 - € 496.00
Specifications
| Antigen | Collagen IV alpha5 |
|---|---|
| Clone | COL4A5/9396R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18699044
|
Novus Biologicals
NBP3-23832-100ug |
100 μg |
€ 496.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678834
|
Novus Biologicals
NBP3-23832-20ug |
20 μg |
€ 279.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Collagen IV alpha5 Monoclonal antibody specifically detects Collagen IV alpha5 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Collagen IV alpha5 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| alpha-5 polypeptide, Alport syndrome, CA54, collagen of basement membrane, alpha-5 chain, collagen, type IV, alpha 5, dA149D17.3, dA24A23.1, MGC167109, MGC42377 | |
| Recombinant fragment (around aa1400-1685) of human Collagen IV alpha5 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| COL4A5/9396R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 1287 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title