missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV Antibody (1043), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97716AF532
This item is not returnable.
View return policy
Description
Collagen IV Monoclonal specifically detects Collagen IV in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.
Specifications
| Collagen IV alpha 1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| COL4A1 | |
| 1043 is a mouse monoclonal IgG1 antibody derived by fusion of Sp 2/0 mouse myeloma cells with spleen cells from a BABL/c mouse immunized with purified human type IV collagen. The immunogen has been isolated from human placenta. | |
| Primary | |
| 1043 recognizes type IV collagen in the extracellular - basement membranes of normal and pathological human tissues. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
| 1043 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting | |
| alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2 | |
| Mouse | |
| 0.1 mL | |
| 1282 | |
| Human, Pig, Bovine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction