missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV Antibody (COL-94), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-22632-0.1ML
This item is not returnable.
View return policy
Description
Collagen IV Monoclonal specifically detects Collagen IV in Human samples. It is validated for Immunohistochemistry.
Specifications
| Collagen IV alpha 1 | |
| Monoclonal | |
| Unconjugated | |
| Ascites, containing less than 0.1% sodium azide with Sodium Azide | |
| COL4A1 | |
| Human collagen type IV. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 1282 | |
| Human | |
| Ascites |
| Immunohistochemistry | |
| COL-94 | |
| Immunohistochemistry | |
| alpha-1 chain, arresten, COL4A1 NC1 domain, collagen alpha-1(IV) chain, collagen IV, alpha-1 polypeptide, collagen, type IV, alpha 1, Collagen-4, EC 3.4.23, EC 6.3.1.2 | |
| Mouse | |
| Ascites | |
| RUO | |
| Primary | |
| This antibody recognizes collagen type IV epitope located on the alpha1 and/or alpha 2 chains of human collagen type IV. This shows no cross reactivity with collagen types I, II, III, V, VI and VII and does not react with human vitronectin, fibronectin or chondroitin sulfate A, B and C. | |
| Store at 4°C. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction