missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen IV Antibody (COL4/8657R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20325-100ug
This item is not returnable.
View return policy
Description
Collagen IV Monoclonal antibody specifically detects Collagen IV in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Collagen IV | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| COL4/8657R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Purified human Collagen IV protein | |
| 100 μg | |
| Cell Biology, Cellular Markers, Extracellular Matrix, Stem Cell Markers | |
| 1282 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction