missing translation for 'onlineSavingsMsg'
Learn More

Collagen VI alpha 1 Antibody [DyLight 550], Novus Biologicals Biologicals™

Product Code. 30517234 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30517234 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30517234 Supplier Novus Biologicals Supplier No. NBP338600R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Collagen VI alpha 1 Polyclonal antibody specifically detects Collagen VI alpha 1 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Collagen VI alpha 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias Collagen 6, collagen alpha-1(VI) chain, collagen VI, alpha-1 polypeptide, collagen, type VI, alpha 1, Collagen-6, human mRNA for collagen VI alpha-1 C-terminal globular domain10alpha 1 (VI) chain (61 AA), OPLL
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 19-250 of human Collagen VI alpha 1 (NP_001839.2).,, Sequence:, AQDEPETPRAVAFQDCPVDLFFVLDTSESVALRLKPYGALVDKVKSFTKRFIDNLRDRYYRCDRNLVWNAGALHYSDEVEIIQGLTRMPGGRDALKSSVDAVKYFGKGTYTDCAIKKGLEQLLVGGSHLKENKYLIVVTDGHPLEGYKEPCGGLEDAVNEAKHLGVKVFSVAITPDHLEPRLSIIATDHTYRRNFTAADWGQSRDAEEAISQTIDTIVDMIKNNVEQVCCSF
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 1291
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.