missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen VI alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15947
This item is not returnable.
View return policy
Description
Collagen VI alpha 1 Polyclonal specifically detects Collagen VI alpha 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Collagen VI | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris-Glycine (pH7), 10% Glycerol with 0.01% Thimerosal | |
| Collagen 6, collagen alpha-1(VI) chain, collagen VI, alpha-1 polypeptide, collagen, type VI, alpha 1, Collagen-6, human mRNA for collagen VI alpha-1 C-terminal globular domain10alpha 1 (VI) chain (61 AA), OPLL | |
| Rabbit | |
| 109 kDa | |
| 0.1 mL | |
| Cytoskeleton Markers, Extracellular Matrix | |
| 1291 | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| P12109 | |
| COL6A1 | |
| Recombinant protein encompassing a sequence within the center region of human COL6A1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse. Expected to cross react based on sequence identity: Bovine (87%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction