missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen VI alpha 3 Antibody (118), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-90328CL1
This item is not returnable.
View return policy
Description
Collagen VI alpha 3 Monoclonal antibody specifically detects Collagen VI alpha 3 in Human samples. It is validated for ELISA
Specifications
| Collagen VI alpha 3 | |
| Monoclonal | |
| CoraFluor 1 | |
| Collagen Alpha-3(VI) Chain, Collagen VI, Alpha-3 Polypeptide, collagen, type VI, alpha 3 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Collagen VI alpha 3 (Uniprot#: P12111-1; 3101T-3177T). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 118 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1293 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction