missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen VI alpha 3 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97126DL594
This item is not returnable.
View return policy
Description
Collagen VI alpha 3 Polyclonal specifically detects Collagen VI alpha 3 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Collagen VI alpha 3 | |
| Polyclonal | |
| Western Blot, ELISA | |
| Collagen Alpha-3(VI) Chain, Collagen VI, Alpha-3 Polypeptide, collagen, type VI, alpha 3 | |
| Produced in rabbits immunized with purified, recombinant Collagen VI alpha 3 (P12111-1; 3101T-3177T). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1293 | |
| Store at 4°C in the dark. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion