missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen VI alpha 3 Rabbit anti-Human, Clone: 118, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 379.00 - € 552.00
Specifications
| Antigen | Collagen VI alpha 3 |
|---|---|
| Clone | 118 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18231176
|
Novus Biologicals
NBP2-90328-50UL |
50 μg |
€ 379.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18267587
|
Novus Biologicals
NBP2-90328-100UL |
100 μg |
€ 552.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Collagen VI alpha 3 Monoclonal specifically detects Collagen VI alpha 3 in Human samples. It is validated for ELISA.Specifications
| Collagen VI alpha 3 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 1293 | |
| IgG | |
| Protein A purified |
| 118 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Collagen Alpha-3(VI) Chain, Collagen VI, Alpha-3 Polypeptide, collagen, type VI, alpha 3 | |
| Recombinant Human Collagen VI alpha 3 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title