missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen VIII alpha 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 292.00 - € 585.00
Specifications
| Antigen | Collagen VIII alpha 1 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18456531
|
Novus Biologicals
NBP2-13856-25ul |
25ul |
€ 292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18799553
|
Novus Biologicals
NBP2-13856 |
0.1 mL |
€ 585.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Collagen VIII alpha 1 Polyclonal specifically detects Collagen VIII alpha 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Collagen VIII alpha 1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 1295 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: QGEYLPDMGLGIDGVKPPHAYGAKKGKNGGPAYEMPAFTAELTAPFPPVGAPVKFNKLLYNGRQNYNPQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| alpha-1 polypeptide, chromosome 3 open reading frame 7, collagen, type VIII, alpha 1, smooth muscle cell-expressed and macrophage conditioned medium-induced proteinsmag-64 | |
| COL8A1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title