missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen XI alpha 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35144-100ul
This item is not returnable.
View return policy
Description
Collagen XI alpha 2 Polyclonal antibody specifically detects Collagen XI alpha 2 in Human samples. It is validated for ELISA,Western Blot
Specifications
| Collagen XI alpha 2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| collagen alpha-2(XI) chain, collagen, type XI, alpha 2, DFNA13, DFNB53, HKE5, PARP, pro-a2 chain of collagen type XI, STL3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 210-380 of human Collagen XI alpha 2 (NP_542411.2).,, Sequence:, IVPGVQAAYESCEQKELECEGGQRERPQNQQPHRAQRSPQQQPSRLHRPQNQEPQSQPTESLYYDYEPPYYDVMTTGTTPDYQDPTPGEEEEILESSLLPPLEEEQTDLQVPPTADRFQAEEYGEGGTDPPEGPYDYTYGYGDDYREETELGPALSAETAHSGAAAHGPRG | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 1302 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction