missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen XXV alpha 1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-289AF594
This item is not returnable.
View return policy
Description
Collagen XXV alpha 1 Polyclonal antibody specifically detects Collagen XXV alpha 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Collagen XXV alpha 1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| Alzheimer disease amyloid-associated protein, AMY, CLAC, CLACP, CLAC-P, Collagen 25, collagen alpha-1(XXV) chain, collagen, type XXV, alpha 1, Collagen-25, collagen-like Alzheimer amyloid plaque component, collagenous Alzheimer amyloid plaque component | |
| A synthetic peptide mapping to residues 430-443 [DYNGNLHEALQRIT] of the NC3 region of human and mouse CLAC-P. | |
| 0.1 mL | |
| Neuroscience | |
| 84570 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction