Learn More
Macherey-Nagel™ Collection Tubes
Product Code. 10083051
Change view
Click to view available options
:
Capacity: 2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10083051 Capacity: 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10083051 Supplier Macherey-Nagel™ Supplier No. 740600
Only null left
Add to basket
Add to basket
Collection Tubes 2 mL, without lid
Specifications
Final Product Type Collection Tubes
Product Type Collection Tubes