Learn More
SCAT Collectors for HPLC
Shop All SCAT Europe Products
Click to view available options
Product Type:
2-in-1 Connector
3-in-1 Connector
3-in-1 Sideways Connector
3-in-1 Vertical Connector
8-in-1 Connector
Diameter (Metric) Outer:
3.2 mm
Unit Size:
Each
Diameter (Metric) Inner:
5.0 to 11.5 mm
Description
For a reliable and clean connection of hoses and capillaries for collecting liquids. SCAT collectors are available in versions with up to 8 connection options.The connections are either on the side or vertically, so that the tubes can be connected easily. Tubes with different diameters from 2.3 to 11.5 mm can be connected. The connectors are made of chemical-resistant PTFE, PFA or PP material.
Order Info
dimForUseWith
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | PFTE, PFA |
| Diameter (Metric) Outer | 3.2 mm |
| For Use With | Tube Connector |
| Shape | Vertical |
| Includes | Fittings for 2 x 2.3/3.2 mm OD, 2-in-1 Connector |
| Color | White |
| Product Type | 2-in-1 Connector |