Learn More
Shamrock™ Color Coded Multi-Purpose Labeling Tape
Offered in a number of colors and sizes to fit your needs. Shamrock™ Color Coded Multi-Purpose Labeling Tape offers multiple opportunities for color coding in the laboratory - test, physician or department.
€ 7.15 - € 148.00
Specifications
Adhesive Type | Removable |
---|---|
Product Type | Color Coded Multi-Purpose Labeling Tape |
Product Code | Brand | Color | Dimensions (LxW) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Dimensions (LxW) | Price | Quantity & Availability | ||||
11731596
|
Shamrock™
ST-11-6 |
Blue | 12.7 m x 38 mm |
€ 17.05
12m |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12320190
|
Shamrock™
ST-10-RNBW |
Assorted | 12.7 m x 25 mm |
€ 124.00
12m |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12310190
|
Shamrock™
ST-12-RNBW |
Assorted | 12.7 m x 12.7 mm |
€ 148.00
Pack of 24 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11701616
|
Shamrock™
ST-20-6 |
Blue | 12.7 m x 50 mm |
€ 19.60
12m |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11700085
|
Shamrock™
ST-10-23 |
Brown | 12.7 m x 25 mm |
€ 12.00
12m |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11867193
|
Shamrock™
ST-12-17 |
Chartreuse | 12.7 m x 12.7 mm |
€ 7.15
12m |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11879070
|
Shamrock™
ST-34-10 |
Copper | 55 m x 19 mm |
€ 10.40
12m |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11827193
|
Shamrock™
ST-12-10 |
Copper | 12.7 m x 12.7 mm |
€ 7.15
12m |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11740075
|
Shamrock™
ST-12-23 |
Dark Brown | 12.7 m x 12.7 mm |
€ 7.15
12m |
In Stock
Log in to see stock. |
||||
11770075
|
Shamrock™
ST-34-23 |
Dark Brown | 55 m x 19 mm |
€ 9.45
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11701596
|
Shamrock™
ST-11-25 |
Green | 12.7 m x 38 mm |
€ 17.02
12m |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11790075
|
Shamrock™
ST-10-14 |
Gray | 12.7 m x 25 mm |
€ 12.00
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11771606
|
Shamrock™
ST-20-3 |
Green | 12.7 m x 50 mm |
€ 19.60
12m |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11721596
|
Shamrock™
ST-11-5 |
Orange | 12.7 m x 38 mm |
€ 17.05
12m |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11791606
|
Shamrock™
ST-20-5 |
Orange | 12.7 m x 50 mm |
€ 19.59
12m |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11887203
|
Shamrock™
ST-34-13 |
Lavender | 55 m x 19 mm |
€ 9.45
12m |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11897203
|
Shamrock™
ST-34-15 |
Lime Green | 55 m x 19 mm |
€ 10.40
12m |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11711616
|
Shamrock™
ST-20-7 |
Pink | 12.7 m x 50 mm |
€ 19.60
12m |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11711596
|
Shamrock™
ST-11-4 |
Red | 12.7 m x 38 mm |
€ 17.05
12m |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11877203
|
Shamrock™
ST-34-11 |
Rose | 55 m x 19 mm |
€ 9.41
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11721606
|
Shamrock™
ST-11-12 |
Violet | 12.7 m x 38 mm |
€ 17.05
12m |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11869070
|
Shamrock™
ST-34-8 |
Tan | 55 m x 19 mm |
€ 10.40
12m |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11791586
|
Shamrock™
ST-11-2 |
Yellow | 12.7 m x 38 mm |
€ 17.02
12m |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11761606
|
Shamrock™
ST-20-2 |
Yellow | 12.7 m x 50 mm |
€ 19.60
12m |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11781586
|
Shamrock™
ST-11-1 |
White | 12.7 m x 38 mm |
€ 17.05
12m |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Resistant to water, oil and acid
- Temperature range of -20 to +121°C
- For autoclave, freezer, refrigerator and oven
- Universal marking with ballpoint pen, paper pencil and felt marker
Specifications
Removable | |
Color Coded Multi-Purpose Labeling Tape |